Learn More
Medchemexpress LLC Phentolamine mesylate | 65-28-1 | 99.9% | 1 ML
SDP
Catalog No. 5000406117
Encompass_Preferred
Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors that expands blood vessels to reduce peripheral vascular resistance. It can be used for the research of pheochromocytoma-related hypertension, heart failure, and erectile dysfunction.
  • Reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors
  • Expands blood vessels to reduce peripheral vascular resistance
  • Used for research of pheochromocytoma-related hypertension
  • Used for research of heart failure
  • Used for research of erectile dysfunction
Catalog No. 50-004-06117 Supplier Medchemexpress LLC Supplier No. HYB0362A10MM/1ML
Phentolamine mesylate (Phentolamine methanesulfonate) is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors that expands blood vessels to reduce peripheral vascular resistance. It can be used for the research of pheochromocytoma-related hypertension, heart failure, and erectile dysfunction.
  • Reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors
  • Expands blood vessels to reduce peripheral vascular resistance
  • Used for research of pheochromocytoma-related hypertension
  • Used for research of heart failure
  • Used for research of erectile dysfunction