Learn More
Medchemexpress LLC Phentolamine mesylate | 65-28-1 | 99.9% | 500 MG
SDP
Catalog No. 5000407278
Encompass_Preferred
Phentolamine mesylate is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors. It expands blood vessels to reduce peripheral vascular resistance and can be used for the research of pheochromocytoma-related hypertension, heart failure, and erectile dysfunction.
  • Reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors
  • Expands blood vessels to reduce peripheral vascular resistance
  • Used for research in pheochromocytoma-related hypertension, heart failure, and erectile dysfunction
  • High purity of 99.9%
Catalog No. 50-004-07278 Supplier Medchemexpress LLC Supplier No. HYB0362A500MG
Phentolamine mesylate is a reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors. It expands blood vessels to reduce peripheral vascular resistance and can be used for the research of pheochromocytoma-related hypertension, heart failure, and erectile dysfunction.
  • Reversible, non-selective, and orally active blocker of α1 and α2 adrenergic receptors
  • Expands blood vessels to reduce peripheral vascular resistance
  • Used for research in pheochromocytoma-related hypertension, heart failure, and erectile dysfunction
  • High purity of 99.9%