Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phentolamine Mesylate, Spectrum™ Chemical
SDP

Catalog No. 18621300 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-300 500 mg
18-621-299 100 mg
18-621-301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-621-300 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1674500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65-28-1

Specifications

Quantity 500 mg
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Phentolamine Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.