Learn More
Cayman Chemical PhentolamInmesylate 100mg
SDP
Catalog No. 502968702
Encompass_Preferred
A reversible antagonist of α-adrenergic receptors, non-specifically binding all α1- and α2-adrenoceptors with nanomolar affinities; rapidly and transiently increases local blood flow
Catalog No. 50-296-8702 Supplier Cayman Chemical Supplier No. 16135100 MG
A reversible antagonist of α-adrenergic receptors, non-specifically binding all α1- and α2-adrenoceptors with nanomolar affinities; rapidly and transiently increases local blood flow