Learn More
Cayman Chemical PhentolamInmesylate 50mg
SDP
Catalog No. 502973213
Encompass_Preferred
A reversible antagonist of α-adrenergic receptors, non-specifically binding all α1- and α2-adrenoceptors with nanomolar affinities; rapidly and transiently increases local blood flow
Catalog No. 50-297-3213 Supplier Cayman Chemical Supplier No. 1613550 MG
A reversible antagonist of α-adrenergic receptors, non-specifically binding all α1- and α2-adrenoceptors with nanomolar affinities; rapidly and transiently increases local blood flow