Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 1-Propenyl Ketone 80.0+%, TCI America™
SDP

Catalog No. P021610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P021610G 10 g
P021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P021610G Supplier TCI America Supplier No. P021610G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 495-41-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00059386
InChI Key FUJZJBCWPIOHHN-QHHAFSJGSA-N
Synonym 1-Phenyl-2-buten-1-one
PubChem CID 231934
IUPAC Name (E)-1-phenylbut-2-en-1-one
SMILES CC=CC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 122°C
Quantity 10 g
Formula Weight 146.19
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl 1-Propenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.