Learn More
Medchemexpress LLC Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate | 108032-93-5 | ≥97.0% | 440.46 g/mol | C20H24O9S | 100 MG
SDP
Catalog No. 5000283185
Encompass_Preferred
Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated thioglycoside reagent used in carbohydrate chemistry and glycosylation research. It functions as an intermediate in organic synthesis and is supplied as a white to off-white solid for laboratory use.
  • Acetylated thioglycoside for glycosylation and carbohydrate synthesis applications.
  • High purity (≥97.0%) suitable for analytical and synthetic work.
  • Provided as a small-scale 100 MG package for bench-top experiments.
  • Solid form simplifies handling and storage.
  • Recommended cold storage to preserve stability and shelf life.
Catalog No. 50-002-83185 Supplier Medchemexpress LLC Supplier No. HYW145527100MG
Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated thioglycoside reagent used in carbohydrate chemistry and glycosylation research. It functions as an intermediate in organic synthesis and is supplied as a white to off-white solid for laboratory use.
  • Acetylated thioglycoside for glycosylation and carbohydrate synthesis applications.
  • High purity (≥97.0%) suitable for analytical and synthetic work.
  • Provided as a small-scale 100 MG package for bench-top experiments.
  • Solid form simplifies handling and storage.
  • Recommended cold storage to preserve stability and shelf life.