Learn More
Medchemexpress LLC Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate | 108032-93-5 | MFCD01862644 | 97.0% | 440.46 g·mol⁻¹ | C20H24O9S | 1 G
SDP
Catalog No. 5000283761
Encompass_Preferred
Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated phenyl thio-glycoside used as a chemical reagent in organic synthesis and carbohydrate research. It is supplied for research use only and is not intended for clinical or human use.
  • Acetylated phenyl thio-glycoside useful for glycosylation and carbohydrate chemistry.
  • Supplied as a pure, characterized material (purity about 97%).
  • Chemical formula C20H24O9S and molecular weight 440.46 g·mol⁻¹.
  • Available in laboratory pack sizes suitable for synthesis and analytical work.
  • Intended for research use only; handle according to standard laboratory safety procedures.
Catalog No. 50-002-83761 Supplier Medchemexpress LLC Supplier No. HYW1455271G
Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated phenyl thio-glycoside used as a chemical reagent in organic synthesis and carbohydrate research. It is supplied for research use only and is not intended for clinical or human use.
  • Acetylated phenyl thio-glycoside useful for glycosylation and carbohydrate chemistry.
  • Supplied as a pure, characterized material (purity about 97%).
  • Chemical formula C20H24O9S and molecular weight 440.46 g·mol⁻¹.
  • Available in laboratory pack sizes suitable for synthesis and analytical work.
  • Intended for research use only; handle according to standard laboratory safety procedures.