Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenyl 1-thio-α-d-mannopyranoside 2,3,4,6-tetraacetate | 108032-93-5 | MFCD01862644 | >97.0% | 440.46 g/mol | C20H24O9S | 250 MG
SDP

Catalog No. 5000282897
Encompass_Preferred

Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated phenyl thio-mannopyranoside provided as a white to off-white solid for research use. It is used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry, glycoside synthesis, and other life-science studies.

  • Acetylated phenyl thio-mannopyranoside structure suitable for glycosylation studies.
  • High purity: >97.0%.
  • Chemical formula: C20H24O9S; molecular weight: 440.46 g/mol.
  • Appearance: white to off-white solid.
  • Available in small laboratory pack sizes, including 100 mg, 250 mg, and 1 g.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months).
  • For research use only.

Catalog No. 50-002-82897 Supplier Medchemexpress LLC Supplier No. HYW145527250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl 1-thio-α-D-mannopyranoside 2,3,4,6-tetraacetate is an acetylated phenyl thio-mannopyranoside provided as a white to off-white solid for research use. It is used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry, glycoside synthesis, and other life-science studies.

  • Acetylated phenyl thio-mannopyranoside structure suitable for glycosylation studies.
  • High purity: >97.0%.
  • Chemical formula: C20H24O9S; molecular weight: 440.46 g/mol.
  • Appearance: white to off-white solid.
  • Available in small laboratory pack sizes, including 100 mg, 250 mg, and 1 g.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.