Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside 98.0+%, TCI America™
SDP

Catalog No. P25215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P25215G Supplier TCI America Supplier No. P25215G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic reagent for biochemical and pharmaceutical research.

Chemical Identifiers

CAS 108032-93-5
Molecular Formula C20H24O9S
Molecular Weight (g/mol) 440.463
MDL Number MFCD01862644
InChI Key JCKOUAWEMPKIAT-SWBPCFCJSA-N
PubChem CID 10993883
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 440.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.