Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 93.0+%, TCI America™
SDP

Catalog No. P2412200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2412200MG 200 mg
P24121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2412200MG Supplier TCI America Supplier No. P2412200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 936326-60-2
Molecular Formula C22H23IO6S
Molecular Weight (g/mol) 542.384
InChI Key GGEMRLZODUITOC-UHFFFAOYSA-M
Synonym Phenyl(2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 131697920
IUPAC Name 4-methylbenzenesulfonate;phenyl-(2,4,6-trimethoxyphenyl)iodanium
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC=CC=C2)OC

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 200 mg
Formula Weight 542.38
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.