Learn More
Medchemexpress LLC Phenyl 2-bromoacetate | 620-72-4 | 99.9% | 215.04 | 1 KG
SDP
Catalog No. 5000404538
Encompass_Preferred
Phenyl 2-bromoacetate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.
  • High purity level
  • Suitable for various life science applications
  • Solid below 31°C, liquid above 33°C
  • Colorless to off-white appearance
  • Soluble in DMSO and SBE-β-CD in saline
  • Pure form storage for up to 3 years at -20°C
  • In-solvent storage for up to 6 months at -80°C
Catalog No. 50-004-04538 Supplier Medchemexpress LLC Supplier No. HY414131KG
Phenyl 2-bromoacetate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only.
  • High purity level
  • Suitable for various life science applications
  • Solid below 31°C, liquid above 33°C
  • Colorless to off-white appearance
  • Soluble in DMSO and SBE-β-CD in saline
  • Pure form storage for up to 3 years at -20°C
  • In-solvent storage for up to 6 months at -80°C