Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 4-Hydroxybenzoate 99.0+%, TCI America™
SDP

Catalog No. H021825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H021825G 25 g
H0218500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H021825G Supplier TCI America Supplier No. H021825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17696-62-7
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00016467
InChI Key GJLNWLVPAHNBQN-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid Phenyl Ester, Phenylparaben
PubChem CID 87250
IUPAC Name phenyl 4-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 182°C
Color White
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.