Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl[4-(trimethylsilyl)thiophen-3-yl]iodonium Trifluoromethanesulfonate 95.0+%, TCI America™
SDP

Catalog No. P2097200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2097200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2097200MG Supplier TCI America Supplier No. P2097200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175224-35-8
Molecular Formula C14H16F3IO3S2Si
Synonym Phenyl[4-(trimethylsilyl)thiophen-3-yl]iodonium Triflate, Phenyl[4-(trimethylsilyl)-3-thienyl]iodonium Trifluoromethanesulfonate, Phenyl[4-(trimethylsilyl)-3-thienyl]iodonium Triflate

Specifications

Melting Point 172°C
Color White
Quantity 200 mg
Formula Weight 508.38
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl[4-(trimethylsilyl)thiophen-3-yl]iodonium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.