Learn More
YMC America, Inc PHENYL 5UM 120A 50X3MM
SDP
Catalog No. 50157066
Encompass_Preferred
PH12S05-0503WT: Phenyl, 5um, 120A, 50X3mm Column
Catalog No. 50-157-066 Supplier YMC America, Inc Supplier No. PH12S050503WT
PH12S05-0503WT: Phenyl, 5um, 120A, 50X3mm Column