Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc PHENYL 5UM 120A 75X30MM
SDP

Catalog No. 50874601
Encompass

Phenyl 5Um; 120A; 75 X 30mm

Catalog No. 50-874-601 Supplier YMC America, Inc Supplier No. PH12S05L530WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl 5Um; 120A; 75 X 30mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.