Learn More
Medchemexpress LLC Phenyl 6-[[3-methoxy-2-[4-(CF3)phenyl]benzoyl]amino]-3,4-dihydro-1H-isoquinoline-2-carboxylate | 913541-47-6 | MFCD28502203 | ≥98.0% | 546.5 g·mol⁻¹ | C31H25F3N2O4 | 100 MG
SDP
Catalog No. 5000244296
Encompass_Preferred
SLX-4090 is a small-molecule, orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor with a reported IC50 of ~8.0 nM. It is used in preclinical research to investigate intestinal lipid absorption and dyslipidemia and is characterized in peer-reviewed literature.
  • Potent MTP inhibition with low-nanomolar IC50.
  • Enterocyte-selective activity targeting intestinal lipid handling.
  • Suitable for preclinical and mechanistic studies of dyslipidemia.
  • High chemical purity for reproducible experimental results.
  • Available as solid and solution formulations for flexibility in assays.
Catalog No. 50-002-44296 Supplier Medchemexpress LLC Supplier No. HY106011100MG
SLX-4090 is a small-molecule, orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor with a reported IC50 of ~8.0 nM. It is used in preclinical research to investigate intestinal lipid absorption and dyslipidemia and is characterized in peer-reviewed literature.
  • Potent MTP inhibition with low-nanomolar IC50.
  • Enterocyte-selective activity targeting intestinal lipid handling.
  • Suitable for preclinical and mechanistic studies of dyslipidemia.
  • High chemical purity for reproducible experimental results.
  • Available as solid and solution formulations for flexibility in assays.