Learn More
Medchemexpress LLC Phenyl 6-[[3-methoxy-2-(4-(trifluoromethyl)phenyl)benzoyl]amino]-3,4-dihydro-1H-isoquinoline | 913541-47-6 | MFCD28502203 | 99.0% | 546.54 g/mol | C31H25F3N2O4 | 25 MG
SDP
Catalog No. 5000243432
Encompass_Preferred
SLX-4090 is an orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor used in preclinical research on dyslipidemia and inhibition of apolipoprotein B secretion. It exhibits potent biochemical activity (IC50 = 8.0 nM) and is supplied as a high-purity research compound.
  • Orally active enterocytic-specific MTP inhibitor.
  • Potent biochemical activity with IC50 8.0 nM.
  • Suitable for dyslipidemia and apolipoprotein B secretion studies.
  • High chemical purity (~98.97%).
  • Molecular formula C31H25F3N2O4 and molecular weight 546.54 g/mol.
  • Available as solid packs and as a 10 mM solution in DMSO.
Catalog No. 50-002-43432 Supplier Medchemexpress LLC Supplier No. HY10601125MG
SLX-4090 is an orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor used in preclinical research on dyslipidemia and inhibition of apolipoprotein B secretion. It exhibits potent biochemical activity (IC50 = 8.0 nM) and is supplied as a high-purity research compound.
  • Orally active enterocytic-specific MTP inhibitor.
  • Potent biochemical activity with IC50 8.0 nM.
  • Suitable for dyslipidemia and apolipoprotein B secretion studies.
  • High chemical purity (~98.97%).
  • Molecular formula C31H25F3N2O4 and molecular weight 546.54 g/mol.
  • Available as solid packs and as a 10 mM solution in DMSO.