Learn More
Medchemexpress LLC Phenyl 6-[[3-methoxy-2-[4-(trifluoromethyl)phenyl]benzoyl]amino]-3,4-dihydro-1H-isoquinoline | 913541-47-6 | MFCD28502203 | 99.0% | 546.54 g/mol | C31H25F3N2O4 | 5 MG
SDP
Catalog No. 5000243076
Encompass_Preferred
SLX-4090 is a potent, orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor used in preclinical research on lipid metabolism and dyslipidemia.
  • Potent MTP inhibitor (IC50 8.0 nM).
  • Inhibits apolipoprotein B secretion in Caco-2 cells; does not inhibit apolipoprotein A1.
  • Orally active in rodents; reduces postprandial lipids with an ED50 of ~7 mg/kg.
  • Molecular weight 546.54 g/mol; chemical formula C31H25F3N2O4.
  • High purity (~98.97% by HPLC).
  • Intended for laboratory research use only; not for therapeutic use.
Catalog No. 50-002-43076 Supplier Medchemexpress LLC Supplier No. HY1060115MG
SLX-4090 is a potent, orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor used in preclinical research on lipid metabolism and dyslipidemia.
  • Potent MTP inhibitor (IC50 8.0 nM).
  • Inhibits apolipoprotein B secretion in Caco-2 cells; does not inhibit apolipoprotein A1.
  • Orally active in rodents; reduces postprandial lipids with an ED50 of ~7 mg/kg.
  • Molecular weight 546.54 g/mol; chemical formula C31H25F3N2O4.
  • High purity (~98.97% by HPLC).
  • Intended for laboratory research use only; not for therapeutic use.