Learn More
Medchemexpress LLC Phenyl 6-(3-methoxy-2-(4-(trifluoromethyl)phenyl)benzoylamino)-3,4-dihydroisoquinoline-2-carboxylate | 913541-47-6 | MFCD28502203 | 99.0% | 546.54 | C31H25F3N2O4 | 50 MG
SDP
Catalog No. 5000244039
Encompass_Preferred
SLX-4090 is an orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor (IC50 8.0 nM) used as a research reagent in dyslipidemia and lipid metabolism studies. It is supplied as a purified solid and is available in solution formats suitable for in vitro and in vivo assays.
  • High potency MTP inhibition (IC50 ~8.0 nM).
  • Enterocytic-specific activity useful for targeted lipid studies.
  • High purity (about 98.97%) for reproducible results.
  • Available as solid and DMSO solution formats for assay flexibility.
  • Suitable for dyslipidemia and lipid metabolism research models.
Catalog No. 50-002-44039 Supplier Medchemexpress LLC Supplier No. HY10601150MG
SLX-4090 is an orally active, enterocytic-specific microsomal triglyceride transfer protein (MTP) inhibitor (IC50 8.0 nM) used as a research reagent in dyslipidemia and lipid metabolism studies. It is supplied as a purified solid and is available in solution formats suitable for in vitro and in vivo assays.
  • High potency MTP inhibition (IC50 ~8.0 nM).
  • Enterocytic-specific activity useful for targeted lipid studies.
  • High purity (about 98.97%) for reproducible results.
  • Available as solid and DMSO solution formats for assay flexibility.
  • Suitable for dyslipidemia and lipid metabolism research models.