Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Acetate, 100g, MP Biomedicals

Catalog No. ICN216573 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216573 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216573 Supplier MP Biomedicals, Inc Supplier No. 0521657380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122-79-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key IPBVNPXQWQGGJP-UHFFFAOYSA-N
Synonym acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech
PubChem CID 31229
ChEBI CHEBI:8082
IUPAC Name phenyl acetate
SMILES CC(=O)OC1=CC=CC=C1

Specifications

Quantity 100 g
Formula Weight 136.14
Chemical Name or Material Phenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.