Learn More
Medchemexpress LLC Phenyl acetate | 122-79-2 | 99.7% | 1 ML
SDP
Catalog No. 5000342918
Encompass_Preferred
Phenyl acetate is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. It plays a role in regulating glutamine metabolism and has been investigated as a potential detection index for certain diseases, including depression. This product is supplied as a high-purity liquid.
  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine
  • Regulates glutamine metabolism
  • Potential detection index for diseases like depression
  • Can block glutaminolysis in lung cancer cell lines in vitro
  • Colorless to light yellow liquid appearance
  • High purity of 99.7%
Catalog No. 50-003-42918 Supplier Medchemexpress LLC Supplier No. HY12873310MM/1ML
Phenyl acetate is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. It plays a role in regulating glutamine metabolism and has been investigated as a potential detection index for certain diseases, including depression. This product is supplied as a high-purity liquid.
  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine
  • Regulates glutamine metabolism
  • Potential detection index for diseases like depression
  • Can block glutaminolysis in lung cancer cell lines in vitro
  • Colorless to light yellow liquid appearance
  • High purity of 99.7%