Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyl acetate

Catalog No. 108723100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-872-3100G 100 g
10-872-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-872-3100G Supplier Sigma Aldrich Supplier No. 108723100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phenyl acetate is an aromatic ester
  • Phenyl acetate levels in urine are marker for the diagnosis of some forms of unipolar major depressive disorders
  • It undergoes fries rearrangement to form a mixture of o- and p-hydroxyacetophenones which are useful intermediates in manufacture of pharmaceuticals

Specifications

CAS 122-79-2
Density 1.073 g/mL (at 25°C (literature))
Boiling Point 196°C (lit.)
Molecular Formula C8H8O2
Linear Formula CH3COOC6H5
Refractive Index n20/D 1.501 (literature)
MDL Number MFCD00008699
Quantity 100 g
Synonym Acetic acid phenyl ester
Molecular Weight (g/mol) 136.15
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.