Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenyl acetate (Standard) | 122-79-2 | 99.7% | 100 MG
SDP

Catalog No. 5000339785
Encompass_Preferred

Phenyl acetate (Standard) is the analytical standard of Phenyl acetate, intended for research and analytical applications. It is identified as an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. This compound plays a role in regulating glutamine metabolism and can serve as a detection index for certain conditions, such as depression.

  • Purity: 99.72%
  • Molecular weight: 136.15
  • Formula: C8H8O2
  • CAS number: 122-79-2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Initial source: Microorganisms, endogenous metabolite
  • Application: Used as an analytical and reference standard for qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.

Catalog No. 50-003-39785 Supplier Medchemexpress LLC Supplier No. HY128733R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl acetate (Standard) is the analytical standard of Phenyl acetate, intended for research and analytical applications. It is identified as an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. This compound plays a role in regulating glutamine metabolism and can serve as a detection index for certain conditions, such as depression.

  • Purity: 99.72%
  • Molecular weight: 136.15
  • Formula: C8H8O2
  • CAS number: 122-79-2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Initial source: Microorganisms, endogenous metabolite
  • Application: Used as an analytical and reference standard for qualitative, quantitative, and methodological research experiments in techniques like HPLC, GC, and MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.