Learn More
Medchemexpress LLC Phenyl acetate (Standard) | 122-79-2 | 99.7% | 250 MG
SDP
Catalog No. 5000342477
Encompass_Preferred
Phenyl acetate (Standard) is an analytical standard and an endogenous metabolite. It is a metabolite of 2-phenylethylamine, found in urine, and regulates glutamine metabolism. It can also serve as a detection index for certain diseases, such as depression. This product is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
  • Endogenous metabolite
  • Can indicate certain diseases like depression
Catalog No. 50-003-42477 Supplier Medchemexpress LLC Supplier No. 250MGHY128733R
Phenyl acetate (Standard) is an analytical standard and an endogenous metabolite. It is a metabolite of 2-phenylethylamine, found in urine, and regulates glutamine metabolism. It can also serve as a detection index for certain diseases, such as depression. This product is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
  • Endogenous metabolite
  • Can indicate certain diseases like depression