Learn More
Medchemexpress LLC Phenyl acetate (Standard) | 122-79-2 | 99.7% | 50 MG
SDP
Catalog No. 5000340741
Encompass_Preferred
Phenyl acetate (Standard) is the analytical standard of Phenyl acetate, intended for research and analytical applications. It is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. Phenyl acetate regulates glutamine metabolism and can serve as a detection index for certain diseases, such as depression.
  • Analytical standard for reference
  • Intended for research and analytical applications
  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine, found in urine
  • Regulates glutamine metabolism
  • Detection index for certain diseases, such as depression
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-40741 Supplier Medchemexpress LLC Supplier No. HY128733R50MG
Phenyl acetate (Standard) is the analytical standard of Phenyl acetate, intended for research and analytical applications. It is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. Phenyl acetate regulates glutamine metabolism and can serve as a detection index for certain diseases, such as depression.
  • Analytical standard for reference
  • Intended for research and analytical applications
  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine, found in urine
  • Regulates glutamine metabolism
  • Detection index for certain diseases, such as depression
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS