Learn More
Medchemexpress LLC Phenyl Agarose (Low Sub) 6FF | 100 ML
SDP
Catalog No. 5000430279
Encompass_Preferred
Phenyl Agarose (Low Sub) 6FF is a low-substitution hydrophobic chromatography medium, suitable for laboratory-scale and industrial-scale purification of biomolecules with relatively weak hydrophobicity. This medium purifies biomolecules based on differences in surface hydrophobicity, where proteins bind in high-salt conditions and are gently eluted by decreasing salt concentration, preserving structure and activity. It is widely used in downstream processing of peptides, antibodies, and protein therapeutics.
  • High mechanical strength
  • Fast flow rate
  • Strong selectivity
  • High recovery of biological activity
  • Environmental safety
  • Resistant to strong acids and bases, allowing for in-place cleaning (CIP) and high-temperature sterilization
  • Lower ligand density and milder elution conditions compared to reverse-phase chromatography media, which better preserve the biological activity of target molecules
Catalog No. 50-004-30279 Supplier Medchemexpress LLC Supplier No. HYK0268100ML
Phenyl Agarose (Low Sub) 6FF is a low-substitution hydrophobic chromatography medium, suitable for laboratory-scale and industrial-scale purification of biomolecules with relatively weak hydrophobicity. This medium purifies biomolecules based on differences in surface hydrophobicity, where proteins bind in high-salt conditions and are gently eluted by decreasing salt concentration, preserving structure and activity. It is widely used in downstream processing of peptides, antibodies, and protein therapeutics.
  • High mechanical strength
  • Fast flow rate
  • Strong selectivity
  • High recovery of biological activity
  • Environmental safety
  • Resistant to strong acids and bases, allowing for in-place cleaning (CIP) and high-temperature sterilization
  • Lower ligand density and milder elution conditions compared to reverse-phase chromatography media, which better preserve the biological activity of target molecules