Learn More
Medchemexpress LLC Phenyl benzoate | 93-99-2 | 99.97% | 198.22 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00982350G
Encompass_Preferred
Phenyl benzoate is a benzoate ester formed by the condensation of phenol with benzoic acid. It acts as a chloride transport blocker, preventing chloride-dependent glutamate accumulation in vesicles. It is also utilized as a preservative in cosmetic products. This product is intended for research use only.
  • Benzoate ester.
  • Chloride transport blocker.
  • Inhibits Cl--dependent Glu accumulation into vesicles.
  • Can be used as a preservative in cosmetic products.
  • For research use only.
Catalog No. 50-004-51615