Learn More
Medchemexpress LLC Phenyl-boronic acid-d5 | 215527-70-1 | MFCD02093711 | 98.0% | 126.96 g/mol | C6H2D5BO2 | 25 G
SDP
Catalog No. 5000208873
Encompass_Preferred
Phenyl-boronic acid-d5 is a deuterium-labeled phenylboronic acid supplied for research use as an isotope-labeled reagent used in analytical chemistry and synthetic applications.
  • Isotope-labeled reagent for analytical and tracer studies.
  • Suitable for mass spectrometry and quantitative analysis.
  • Deuterium-labeled phenyl ring (d5) for minimal isotope scrambling.
  • Available in small-scale research pack sizes (pack size varies by supplier).
Catalog No. 50-002-08873 Supplier Medchemexpress LLC Supplier No. 25GHYW067615
Phenyl-boronic acid-d5 is a deuterium-labeled phenylboronic acid supplied for research use as an isotope-labeled reagent used in analytical chemistry and synthetic applications.
  • Isotope-labeled reagent for analytical and tracer studies.
  • Suitable for mass spectrometry and quantitative analysis.
  • Deuterium-labeled phenyl ring (d5) for minimal isotope scrambling.
  • Available in small-scale research pack sizes (pack size varies by supplier).