Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyl chloroformate

Catalog No. 237906100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-790-6100G 100 g
23-790-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-790-6100G Supplier Sigma Aldrich Supplier No. 237906100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Kinetics of spontaneous hydrolysis of phenyl chloroformate has been studied in water-ethylene glycol, cationic, zwitterionic, nonionic and anionic micellar solutions.Phenyl chloroformate has been used in the synthesis of poly(2-(phenoxycarbonyloxy)ethylmethacrylate) and phenyl-(4-vinylphenyl)carbonate.

Specifications

CAS 1885-14-9
Density 1.248 g/mL (at 25°C (literature))
Boiling Point 74°C to 75°C (13 mmHg)
Molecular Formula C7H5ClO2
Refractive Index n20/D 1.511 (literature)
Linear Formula ClCOOC6H5
MDL Number MFCD00000637
Quantity 100 g
Synonym Chloroformic acid phenyl ester
Molecular Weight (g/mol) 156.57
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.