Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl chlorothionocarbonate, 99%

Catalog No. p-4509973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215490010 1 g Glass bottle
AC215490050 5 g Glass bottle
AC215490250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC215490010 Supplier Thermo Scientific Chemicals Supplier No. 215490010
Only null left
Add to Cart

CAS: 1005-56-7 | C7H5ClOS | 172.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005-56-7
Molecular Formula C7H5ClOS
Molecular Weight (g/mol) 172.63
MDL Number MFCD00004920
InChI Key KOSYAAIZOGNATQ-UHFFFAOYSA-N
Synonym o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate
PubChem CID 70498
IUPAC Name O-phenyl chloromethanethioate
SMILES ClC(=S)OC1=CC=CC=C1

Specifications

Color Yellow
Density 1.2480g/mL
Boiling Point 81.0°C to 83.0°C (6.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4795 to 1.4815
Linear Formula ClC(S)OC6H5
Quantity 1 g
Beilstein 06,III,609
Fieser 10,306
Specific Gravity 1.248
Solubility Information Solubility in water: decomposes
Formula Weight 172.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl chlorothionocarbonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 213-736-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.