Learn More
Cayman Chemical Phenyl Cyclopentyl Ketone, Purity: ≥98%, CAS: 5422-88-8, 10 mg
SDP
Catalog No. 502989795
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of ketamine; intended for research and forensic applications
Catalog No. 50-298-9795 Supplier Cayman Chemical Supplier No. 3103610 MG
An analytical reference standard categorized as a precursor in the synthesis of ketamine; intended for research and forensic applications