Learn More
Medchemexpress LLC Phenyl β-D-glucopyranoside | 1464-44-4 | 99.85% | 256.25 | 500 MG
SDP
Catalog No. 5000453746
Encompass_Preferred
Phenyl β-D-glucopyranoside is a component isolated from Phellodendron amurense, which exhibits anti-inflammatory and anti-tumor activities. It inhibits nitric oxide (NO) production and the expression of iNOS and COX-2. Additionally, it inhibits the nuclear translocation of NF-κB and the expression of pro-inflammatory cytokines and related genes.
  • Isolated from Phellodendron amurense.
  • Anti-inflammatory activity.
  • Anti-tumor activity.
  • Inhibits nitric oxide (NO) production.
  • Inhibits iNOS and COX-2 expression.
  • Inhibits nuclear translocation of NF-κB.
  • Inhibits expression of pro-inflammatory cytokines and related genes.
Catalog No. 50-004-53746 Supplier Medchemexpress LLC Supplier No. HYW011849500MG
Phenyl β-D-glucopyranoside is a component isolated from Phellodendron amurense, which exhibits anti-inflammatory and anti-tumor activities. It inhibits nitric oxide (NO) production and the expression of iNOS and COX-2. Additionally, it inhibits the nuclear translocation of NF-κB and the expression of pro-inflammatory cytokines and related genes.
  • Isolated from Phellodendron amurense.
  • Anti-inflammatory activity.
  • Anti-tumor activity.
  • Inhibits nitric oxide (NO) production.
  • Inhibits iNOS and COX-2 expression.
  • Inhibits nuclear translocation of NF-κB.
  • Inhibits expression of pro-inflammatory cytokines and related genes.