Learn More
Medchemexpress LLC Phenyl β-D-glucopyranoside | 1464-44-4 | 99.9% | 256.25 | 5 G
SDP
Catalog No. 5000453820
Encompass_Preferred
Phenyl β-D-glucopyranoside is a component isolated from *Phellodendron amurense* that exhibits anti-inflammatory and anti-tumor activities. It inhibits nitric oxide (NO) production, and the expression of iNOS and COX-2. It also inhibits the nuclear translocation of NF-κB and the expression of pro-inflammatory cytokines and related genes.
  • Component isolated from *Phellodendron amurense*
  • Anti-inflammatory activity
  • Anti-tumor activity
  • Inhibits nitric oxide (NO) production
  • Inhibits expression of iNOS and COX-2
  • Inhibits nuclear translocation of NF-κB
  • Inhibits expression of pro-inflammatory cytokines and related genes
Catalog No. 50-004-53820 Supplier Medchemexpress LLC Supplier No. HYW0118495G
Phenyl β-D-glucopyranoside is a component isolated from *Phellodendron amurense* that exhibits anti-inflammatory and anti-tumor activities. It inhibits nitric oxide (NO) production, and the expression of iNOS and COX-2. It also inhibits the nuclear translocation of NF-κB and the expression of pro-inflammatory cytokines and related genes.
  • Component isolated from *Phellodendron amurense*
  • Anti-inflammatory activity
  • Anti-tumor activity
  • Inhibits nitric oxide (NO) production
  • Inhibits expression of iNOS and COX-2
  • Inhibits nuclear translocation of NF-κB
  • Inhibits expression of pro-inflammatory cytokines and related genes