Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl β -D-glucopyranoside 98%, Thermo Scientific™

Catalog No. AC270820050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC270820050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC270820050 Supplier Thermo Scientific Chemicals Supplier No. 270820050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1464-44-4
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.25
MDL Number MFCD03410292
InChI Key NEZJDVYDSZTRFS-WLEIBRHLNA-N
Synonym phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d
PubChem CID 65080
ChEBI CHEBI:28749
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 172°C to 176°C
Color White to Beige
Infrared Spectrum Authentic
Quantity 5 g
Beilstein 31, 205
Specific Rotation -69° to -72° (20°C, 589nm) (c=1, H2O)
Formula Weight 256.26
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Phenyl β -D-glucopyranoside, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.