Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl-β-D-glucoside, MP Biomedicals™

Catalog No. ICN10489405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10489405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10489405 Supplier MP Biomedicals, Inc Supplier No. 0210489405

missing translation for 'validMainframeMsg'

Specifications

CAS 1464-44-4
Molecular Formula C12H16O6
MDL Number MFCD03410292
Quantity 5 g
Synonym phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d
InChI Key NEZJDVYDSZTRFS-WLEIBRHLNA-N
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 256.25
PubChem CID 65080
ChEBI CHEBI:28749
Formula Weight 256.3
Physical Form Crystalline
Chemical Name or Material Phenyl-β-D-glucoside
Show More Show Less
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.