Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyl dichlorophosphate

Catalog No. P2238925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2238925G 25 g
P22389100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2238925G Supplier Sigma Aldrich Supplier No. P2238925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the preparation of phosphate diesters.

Specifications

CAS 770-12-7
Density 1.412 g/mL (at 25°C (literature))
Boiling Point 241°C to 243°C (lit.)
Molecular Formula C6H5Cl2O2P
Refractive Index n20/D 1.523 (literature)
Linear Formula C6H5OP(O)Cl2
MDL Number MFCD00002067
Quantity 25 g
Synonym Phenyl phosphodichloridate; Phenyl phosphorodichloridate; Phenyl phosphoryl dichloride
Molecular Weight (g/mol) 210.98
Percent Purity ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.