Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl disulfide, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
882-33-7
C12H10S2
218.33
MFCD00003065
GUUVPOWQJOLRAS-UHFFFAOYSA-N
diphenyl disulfide,phenyl disulfide,disulfide, diphenyl,diphenyldisulfide,1,2-diphenyldisulfane,phenyldisulfanyl benzene,diphenyl disulphide,biphenyl disulfide,disulfide diphenyl,phenyldithiobenzene
13436
(phenyldisulfanyl)benzene
C1=CC=C(C=C1)SSC2=CC=CC=C2
882-33-7
C12H10S2
218.33
MFCD00003065
GUUVPOWQJOLRAS-UHFFFAOYSA-N
diphenyl disulfide,phenyl disulfide,disulfide, diphenyl,diphenyldisulfide,1,2-diphenyldisulfane,phenyldisulfanyl benzene,diphenyl disulphide,biphenyl disulfide,disulfide diphenyl,phenyldithiobenzene
13436
(phenyldisulfanyl)benzene
C1=CC=C(C=C1)SSC2=CC=CC=C2
Boiling Point 191°C to 192°C (15.0mmHg)
Infrared Spectrum Authentic
Quantity 50g
Color White
Packaging Plastic bottle
Melting Point 58°C to 61°C
Assay Percent Range 98.5% min. (HPLC)
Linear Formula (C6H5S-)2
Beilstein 06, 323
Fieser 05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156
Merck Index 15, 3355
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 218.33
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material Phenyl disulfide, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 212-926-4

RTECSNumber : SS6825000

TSCA : TSCA