Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Ether-biphenyl Eutectic

Catalog No. AC417520010
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC417520010 1 kg
AC417521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417520010 Supplier Thermo Scientific Chemicals Supplier No. 417520010
Add to Cart

CAS: 8004-13-5 | C24H20O | 324.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-13-5
Molecular Formula C24H20O
Molecular Weight (g/mol) 324.41
MDL Number MFCD00148859
InChI Key MHCVCKDNQYMGEX-UHFFFAOYSA-N
Synonym diphyl, dowtherm, dowtherm a, dinil, dinyl, therminol vp, phenyl ether-biphenyl mixture, phenyl ether-diphenyl mixture, biphenyl-diphenyl ether mixture, hsdb 137
PubChem CID 24670
IUPAC Name 1,1'-biphenyl;phenoxybenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2.C1=CC=C(C=C1)OC2=CC=CC=C2

Specifications

Melting Point 13°C
Density 1.0560g/mL
Flash Point 123°C
Packaging Glass Bottle
Refractive Index 1.5885 to 1.5905
Quantity 1 kg
Specific Gravity 1.056
Formula Weight 324.41
Chemical Name or Material Phenyl ether-biphenyl eutectic
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
RTECSNumber DV1500000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.