Learn More
Cayman Chemical Phenyl fentnyld5hydroCLO 100ug
SDP
Catalog No. 502919058
Encompass_Preferred
An internal standard for the quantification of phenyl fentanyl by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-291-9058 Supplier Cayman Chemical Supplier No. 39197100 UG
An internal standard for the quantification of phenyl fentanyl by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications