Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl formate, 95%

Catalog No. AAL0028906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0028906 5 g
AAL0028914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0028906 Supplier Thermo Scientific Chemicals Supplier No. L0028906
Add to Cart

CAS: 1864-94-4 | C7H6O2 | 122.123 g/mol

Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl formate is widely used in the palladium-catalyzed carbonylation of aryl, alkenyl and allyl halides. It is also used as a reagent for the formylation of amines.

Solubility
Immiscible with water, alcohol, ether and benzene.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1864-94-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00014126
InChI Key GEOWCLRLLWTHDN-UHFFFAOYSA-N
Synonym formic acid phenyl ester, formic acid, phenyl ester, phocho, formic acid, phenylester, acmc-1box2, phenyl formate gc
PubChem CID 74626
IUPAC Name phenyl formate
SMILES C1=CC=C(C=C1)OC=O

Specifications

Density 1.09
Boiling Point 72°C to 74°C (13 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.512
Quantity 5 g
Beilstein 507113
Solubility Information Immiscible with water,alcohol,ether and benzene.
Formula Weight 122.12
Percent Purity 95%
Chemical Name or Material Phenyl formate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-471-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.