Learn More
Waters Corp CSH Phenyl-Hexyl Column, VanGuard FIT, 1.7 μm, 2.1 × 50 mm
Catalog No. NC3385879 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH Phenyl-Hexyl Columns provide alternative analyte selectivity, making them a valuable tool for method development. Featuring MaxPeak High Performance Surface hardware and integrated VanGuard FIT design, these columns reduce unwanted analyte/surface interactions and extend operational lifetime.
  • Trifunctionally bonded C6 phenyl ligand for robust, low bleed performance and selective retention of polyaromatic compounds
  • Offers complementary selectivity to straight-chain alkyl phases
  • CSH particle morphology ensures excellent peak shape and improved sample loadability for basic compounds
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Supports simpler mobile phases, reducing method development complexity
  • Excellent MS performance with formic acid modifier
  • Enables fast pH switching and column equilibration
Catalog No. NC3385879 Supplier Waters Corp Supplier No. 186009477
ACQUITY Premier CSH Phenyl-Hexyl Columns provide alternative analyte selectivity, making them a valuable tool for method development. Featuring MaxPeak High Performance Surface hardware and integrated VanGuard FIT design, these columns reduce unwanted analyte/surface interactions and extend operational lifetime.
  • Trifunctionally bonded C6 phenyl ligand for robust, low bleed performance and selective retention of polyaromatic compounds
  • Offers complementary selectivity to straight-chain alkyl phases
  • CSH particle morphology ensures excellent peak shape and improved sample loadability for basic compounds
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Supports simpler mobile phases, reducing method development complexity
  • Excellent MS performance with formic acid modifier
  • Enables fast pH switching and column equilibration