Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenyl hydrogen sulfate | 937-34-8 | MFCD00059840 | 99.6% | 174.17 | C6H6O4S | 5 MG
SDP

Catalog No. 5000264119
Encompass_Preferred

Phenyl sulfate is an analytical standard and a gut microbiota-derived metabolite used in biochemical and metabolic research. It is associated with albuminuria and podocyte damage in experimental models and is used as a marker in studies of diabetic kidney disease.

  • Analytical standard for biochemical and metabolic studies.
  • Molecular formula: C6H6O4S.
  • Molecular weight: 174.17 g/mol.
  • High purity (~99.6%) suitable for analytical applications.
  • Available as solid (1, 5, 10, 25, 50, 100 mg) and solution (10 mM, 1 mL).
  • Physical state: solid; also supplied as aqueous solution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Used as a marker in renal disease and diabetic kidney disease research.

Catalog No. 50-002-64119 Supplier Medchemexpress LLC Supplier No. HY1284425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl sulfate is an analytical standard and a gut microbiota-derived metabolite used in biochemical and metabolic research. It is associated with albuminuria and podocyte damage in experimental models and is used as a marker in studies of diabetic kidney disease.

  • Analytical standard for biochemical and metabolic studies.
  • Molecular formula: C6H6O4S.
  • Molecular weight: 174.17 g/mol.
  • High purity (~99.6%) suitable for analytical applications.
  • Available as solid (1, 5, 10, 25, 50, 100 mg) and solution (10 mM, 1 mL).
  • Physical state: solid; also supplied as aqueous solution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Used as a marker in renal disease and diabetic kidney disease research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.