Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyl Isothiocyanate

Catalog No. 3178611G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-786-11G 1 g
31-786-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-786-11G Supplier Sigma Aldrich Supplier No. 3178611G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenyl Isothiocyanate

Specifications

CAS 103-72-0
Melting Point -21°C
Density 1.132 g/mL at 20°C
Boiling Point 218°C; 221°C
Molecular Formula C7H5NS
Refractive Index n20/D 1.6515
Linear Formula C6H5NCS
MDL Number MFCD00004798
Quantity 1 g
Synonym PITC
Molecular Weight (g/mol) 135.19 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.