Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenyl isothiocyanate for
Catalog No. 502898219
Encompass
Phenyl isothiocyanate is an aromatic isothiocyanate. It participates in dehydration reactions of alcohols. It is widely used for synthesis of various biologically important heterocyclic compounds.
Catalog No. 50-289-8219 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7878025ML
Phenyl isothiocyanate is an aromatic isothiocyanate. It participates in dehydration reactions of alcohols. It is widely used for synthesis of various biologically important heterocyclic compounds.