Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Malonic Acid, MP Biomedicals

Catalog No. ICN211394 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211394 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211394 Supplier MP Biomedicals, Inc Supplier No. 0521139405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2613-89-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00004253
InChI Key WWYDYZMNFQIYPT-UHFFFAOYSA-N
Synonym phenylmalonic acid, 2-phenylmalonic acid, propanedioic acid, phenyl, phenylpropanedioc acid, phenylmalonicacid, malonic acid, phenyl, chembl78794, propanedioic acid, 2-phenyl, .alpha.-carboxyphenylacetic acid, toluene-.alpha.,.alpha.-dicarboxylic acid
PubChem CID 75791
IUPAC Name 2-phenylpropanedioic acid
SMILES OC(=O)C(C(O)=O)C1=CC=CC=C1

Specifications

Quantity 10 g
Formula Weight 180.16
Chemical Name or Material Phenyl Malonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.