Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g/mol | C10H10O2 | 10 G
SDP
Catalog No. 5000301508
Encompass_Preferred
Phenyl methacrylate is a hydrophobic methacrylate monomer (CAS 2177-70-0) used in polymer synthesis and materials research, including reported use in preparing light-defocusing plastic rods. It is supplied as a liquid reagent and should be handled under inert conditions and protected from light to minimize polymerization and degradation.
  • Hydrophobic monomer suitable for polymer synthesis.
  • Useful for specialty applications such as light-defocusing plastic rods.
  • Liquid reagent with a reported density of 1.053 g/cm³.
  • Store under inert atmosphere and protect from light to preserve quality.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-003-01508 Supplier Medchemexpress LLC Supplier No. HYW01721310G
Phenyl methacrylate is a hydrophobic methacrylate monomer (CAS 2177-70-0) used in polymer synthesis and materials research, including reported use in preparing light-defocusing plastic rods. It is supplied as a liquid reagent and should be handled under inert conditions and protected from light to minimize polymerization and degradation.
  • Hydrophobic monomer suitable for polymer synthesis.
  • Useful for specialty applications such as light-defocusing plastic rods.
  • Liquid reagent with a reported density of 1.053 g/cm³.
  • Store under inert atmosphere and protect from light to preserve quality.
  • Available in small laboratory pack sizes for research use.