Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g/mol | C10H10O2 | 100 G
SDP
Catalog No. 5000300347
Encompass_Preferred
Phenyl methacrylate is a hydrophobic methacrylate monomer (C10H10O2, MW 162.19 g/mol, CAS 2177-70-0) used in polymer synthesis and specialty material applications such as light-defocusing plastic rods (LDR). Supplied as a liquid, it exhibits a density of about 1.053 g/cm³ and should be protected from light and stored under an inert atmosphere.
  • Hydrophobic methacrylate monomer suitable for radical polymerization.
  • Introduces phenyl functionality for increased rigidity and chemical resistance.
  • Useful in specialty polymers and optical materials such as light-defocusing rods.
  • Liquid form with reported density ~1.053 g/cm³ for formulation calculations.
  • Recommended storage under inert atmosphere and protection from light to preserve stability.
Catalog No. 50-003-00347 Supplier Medchemexpress LLC Supplier No. HYW017213100G
Phenyl methacrylate is a hydrophobic methacrylate monomer (C10H10O2, MW 162.19 g/mol, CAS 2177-70-0) used in polymer synthesis and specialty material applications such as light-defocusing plastic rods (LDR). Supplied as a liquid, it exhibits a density of about 1.053 g/cm³ and should be protected from light and stored under an inert atmosphere.
  • Hydrophobic methacrylate monomer suitable for radical polymerization.
  • Introduces phenyl functionality for increased rigidity and chemical resistance.
  • Useful in specialty polymers and optical materials such as light-defocusing rods.
  • Liquid form with reported density ~1.053 g/cm³ for formulation calculations.
  • Recommended storage under inert atmosphere and protection from light to preserve stability.