Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g/mol | C10H10O2 | 25 G
SDP
Catalog No. 5000304211
Encompass_Preferred
Phenyl methacrylate is a hydrophobic methacrylate monomer (the phenyl ester of methacrylic acid) commonly used in polymer synthesis and specialty materials research. It is provided as a stabilized liquid reagent (C10H10O2, 162.19 g/mol) for laboratory-scale formulation and material fabrication.
  • Hydrophobic methacrylate monomer suitable for copolymerization.
  • Useful for synthesis of light-defocusing plastic rods (LDR).
  • Liquid form allows accurate dosing and easy mixing into formulations.
  • Known molecular formula and molecular weight support reproducible formulation.
  • Supplied with documentation for safe laboratory handling.
Catalog No. 50-003-04211 Supplier Medchemexpress LLC Supplier No. 25GHYW017213
Phenyl methacrylate is a hydrophobic methacrylate monomer (the phenyl ester of methacrylic acid) commonly used in polymer synthesis and specialty materials research. It is provided as a stabilized liquid reagent (C10H10O2, 162.19 g/mol) for laboratory-scale formulation and material fabrication.
  • Hydrophobic methacrylate monomer suitable for copolymerization.
  • Useful for synthesis of light-defocusing plastic rods (LDR).
  • Liquid form allows accurate dosing and easy mixing into formulations.
  • Known molecular formula and molecular weight support reproducible formulation.
  • Supplied with documentation for safe laboratory handling.