Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g·mol⁻¹ | C10H10O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0172135G
Encompass_Preferred
Phenyl methacrylate is an aromatic methacrylate monomer (C10H10O2, CAS 2177-70-0) used to introduce hydrophobic phenyl ester functionality into polymers and copolymers. It is applied in material and polymer synthesis, including reported use in light-defocusing plastic rod (LDR) fabrication, and is polymerizable under typical methacrylate reaction conditions.
  • Hydrophobic monomer with aromatic phenyl ester functionality.
  • Suitable for free-radical polymerization and copolymerization.
  • Useful for specialty resins and optical material applications such as LDRs.
  • Compatible with standard methacrylate polymerization conditions.
  • Available in multiple research and bulk pack sizes.
Catalog No. 50-003-01511